Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Diazo-4'-methoxydiphenylamine Sulfate 98.0+%, TCI America™
SDP

Catalog No. D229025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D229025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D229025G Supplier TCI America Supplier No. D229025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49732-38-9
Molecular Formula C13H13N3O5S
Molecular Weight (g/mol) 323.323
MDL Number MFCD00080910
InChI Key UXVMTJSLLAUFTO-UHFFFAOYSA-M
Synonym Azoic Diazo Component 35, 4-[N-(4-Methoxyphenyl)amino]benzenediazonium Sulfate
PubChem CID 44630256
IUPAC Name hydrogen sulfate;4-(4-methoxyanilino)benzenediazonium
SMILES COC1=CC=C(C=C1)NC2=CC=C(C=C2)[N+]#N.OS(=O)(=O)[O-]

Specifications

Quantity 25 g
Formula Weight 323.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Diazo-4′-methoxydiphenylamine Sulfate
EINECSNumber (5)-2320
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.