CAS RN 49757-42-8
IUPAC Name:
1-[chlorobis(4-methoxyphenyl)methyl]-4-methoxybenzene
Synonyms:
1-[chlorobis(4-methoxyphenyl)methyl]-4-methoxybenzene
Molecular Weight (g/mol):
368.86
Molecular Formula:
C22H21ClO3
InChi Key:
OBFCMKPFILBCSQ-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C(Cl)(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
Sigma Aldrich Chlorotris(Triphenylphosphine)Cobalt(I)
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.