accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SSNHO

CAS RN 49762-08-5

IUPAC Name: 3-hydroxy-4-methyl-2,3-dihydro-1,3-thiazole-2-thione
Synonyms: 3-hydroxy-4-methyl-1,3-thiazole-2-thione
Molecular Weight (g/mol): 147.21
Molecular Formula: C4H5NOS2
InChi Key: LOKDIDVKVVVVHK-UHFFFAOYSA-N
SMILES: CC1=CSC(=S)N1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results