Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4-methyl-2(3H)-thiazolethione 98.0+%, TCI America™
SDP

Catalog No. H09021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09021G Supplier TCI America Supplier No. H09021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49762-08-5
Molecular Formula C4H5NOS2
Molecular Weight (g/mol) 147.21
MDL Number MFCD00077793
InChI Key LOKDIDVKVVVVHK-UHFFFAOYSA-N
Synonym 2-Mercapto-4-methylthiazole 3-Oxide
PubChem CID 341593
IUPAC Name 3-hydroxy-4-methyl-1,3-thiazole-2-thione
SMILES CC1=CSC(=S)N1O

Specifications

Melting Point 96°C
Quantity 1 g
Formula Weight 147.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4-methyl-2(3H)-thiazolethione [for Source of Alkyl Radical]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.