accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN(R)OH(S)(S)O(R)

CAS RN 498-45-3

IUPAC Name: (1R,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-ol
Synonyms: scopine
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: FIMXSEMBHGTNKT-UPGAHCIJSA-N
SMILES: CN1[C@H]2CC(O)C[C@@H]1[C@H]1O[C@@H]21
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results