Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scopine 98.0+%, TCI America™
SDP

Catalog No. S0912200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0912200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S0912200MG Supplier TCI America Supplier No. S0912200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 498-45-3
Molecular Formula C8H13NO2
Molecular Weight (g/mol) 155.20
MDL Number MFCD06658282
InChI Key FIMXSEMBHGTNKT-UPGAHCIJSA-N
Synonym 6,7-Epoxytropine
PubChem CID 1274465
IUPAC Name (1R,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-ol
SMILES CN1[C@H]2CC(O)C[C@@H]1[C@H]1O[C@@H]21

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 200 mg
Formula Weight 155.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Scopine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.