accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOHN

CAS RN 498-63-5

IUPAC Name: (pyrrolidin-2-yl)methanol
Synonyms: 2-pyrrolidinemethanol
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: HVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILES: OCC1CCCN1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)