accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COOOOCH2OOCH2OOCH2

CAS RN 4986-89-4

IUPAC Name: 3-(prop-2-enoyloxy)-2,2-bis[(prop-2-enoyloxy)methyl]propyl prop-2-enoate
Synonyms: pentaerythritol tetraacrylate
Molecular Weight (g/mol): 352.34
Molecular Formula: C17H20O8
InChi Key: KNSXNCFKSZZHEA-UHFFFAOYSA-N
SMILES: C=CC(=O)OCC(COC(=O)C=C)(COC(=O)C=C)COC(=O)C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results