Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pentaerythritol tetraacrylate

Catalog No. 408263250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-826-3250ML 250 mL
40-826-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 40-826-3250ML Supplier Sigma Aldrich Supplier No. 408263250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Pentaerythritol tetraacrylate (PETA) is a hydrophobic tetrafunctional monomer that is majorly used as a crosslinking agent that facilitates an improvement in the surface morphology of the polymeric material. It can be synthesized by the suspension of pentaerythritol in the solution of triethylamine and dichloromethane.PETA can be used in the synthesis of gel polymer electrolyte for the fabrication of lithium-sulfur batteries.

Specifications

CAS 4986-89-4
Density 1.19 g/mL (at 25°C)
Molecular Formula C17H20O8
Linear Formula (H2C=CHCO2CH2)4 C
Refractive Index n20/D 1.487 (literature)
MDL Number MFCD00015334
Quantity 250 mL
Synonym 2,2-Bis[[(1-oxo-2-propenyl)oxy]methyl]-1,3-propanediyl ester; Tetramethylolmethane tetraacrylate
Molecular Weight (g/mol) 352.34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.