accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FNOO

CAS RN 499-08-1

IUPAC Name: 1-fluoro-3-methyl-5-nitrobenzene
Synonyms: 1-fluoro-3-methyl-5-nitrobenzene
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: IPMURRZVEWZEPP-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(F)=C1)[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results