accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3HO

CAS RN 499-75-2

IUPAC Name: 2-methyl-5-(propan-2-yl)phenol
Synonyms: carvacrol caswell no. 511 carvacrol (substance) epa pesticide chemical code 022104 isopropyl-o-cresol o-thymol
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: RECUKUPTGUEGMW-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C)C(O)=C1
Boiling Point 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)