Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carvacrol, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018971 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8971 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8971 Supplier MilliporeSigma Supelco Supplier No. 4263250MG
Add to Cart
Add to Cart

Carvacrol is a monoterpenic phenol, having a pungent type odor. It is a major constituent, which is commonly present in essential oil fractions of oregano and thyme, and can play the role of an antimicrobial agent.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 499-75-2
Density 0.976 g/mL (at 20°C)
Flash Point 106°C
Physical Form Neat
Linear Formula (CH3)2 CHC6H3(CH3)OH
MDL Number MFCD00002236
Melting Point 3°C to 4°C
Boiling Point 236°C to 237°C
Synonym 5-Isopropyl-2-methylphenol
Type Analytical Standard
Quantity 50 mg
Formula Weight 150.22
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.