accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 499770-76-2

H3CNBrNN

CAS RN 499770-76-2

IUPAC Name: 5-(bromomethyl)-1-methyl-1H-1,2,3-benzotriazole
Synonyms: 5-(bromomethyl)-1-methyl-1,2,3-benzotriazole
Molecular Weight (g/mol): 226.08
Molecular Formula: C8H8BrN3
InChi Key: OSUZHHPMRAIJDY-UHFFFAOYSA-N
SMILES: CN1N=NC2=CC(CBr)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results