Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Bromomethyl)-1-methyl-1H-1,2,3-benzotriazole, 90+%, Thermo Scientific™

Catalog No. CC23508DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC23508DE 5 g Amber Glass Bottle
CC23508CB 250 mg Amber Glass Bottle
CC23508DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC23508DE Supplier Thermo Scientific Chemicals Supplier No. CC23508DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 499770-76-2
Molecular Formula C8H8BrN3
Molecular Weight (g/mol) 226.077
InChI Key OSUZHHPMRAIJDY-UHFFFAOYSA-N
Synonym 5-bromomethyl-1-methyl-1h-benzo d 1,2,3 triazole, 5-bromomethyl-1-methyl-1h-1,2,3-benzotriazole, 5-bromomethyl-1-methylbenzotriazole, 5-bromomethyl-1-methyl-benzotriazole, 1h-benzotriazole, 5-bromomethyl-1-methyl, 5-bromomethyl-1-methyl-1,2,3-benzotriazole, 5-bromomethyl-1-methyl-1h-benzotriazole
PubChem CID 2795418
IUPAC Name 5-(bromomethyl)-1-methylbenzotriazole
SMILES CN1C2=C(C=C(C=C2)CBr)N=N1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 226.08
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-1-methyl-1H-1, 2, 3-benzotriazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.