accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNFFFN

CAS RN 499796-94-0

IUPAC Name: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
Synonyms: 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
Molecular Weight (g/mol): 268.26
Molecular Formula: C12H7F3N2S
InChi Key: GTHUVDGNMKVNDQ-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=C(C=C1)C(F)(F)F)C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results