Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile, 97%, Thermo Scientific™

Catalog No. MO00862DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00862DA 1 g Amber Glass Bottle
MO00862CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO00862DA Supplier Thermo Scientific Chemicals Supplier No. MO00862DA

Chemical Identifiers

CAS 499796-94-0
Molecular Formula C12H7F3N2S
Molecular Weight (g/mol) 268.257
InChI Key GTHUVDGNMKVNDQ-UHFFFAOYSA-N
Synonym 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carbonitrile, 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carbonitrile
PubChem CID 2794633
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carbonitrile
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 268.26
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.