CAS RN 4998-57-6
IUPAC Name:
2-amino-3-(1H-imidazol-5-yl)propanoic acid
Synonyms:
histidine
Molecular Weight (g/mol):
155.16
Molecular Formula:
C6H9N3O2
InChi Key:
HNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILES:
NC(CC1=CN=CN1)C(O)=O
1
–
1
of
1
results
1
DL-Histidine 98.0+%, TCI America™
CAS: 4998-57-6 Molecular Formula: C6H9N3O2 Molecular Weight (g/mol): 155.157 MDL Number: MFCD00005208 InChI Key: HNDVDQJCIGZPNO-UHFFFAOYSA-N Synonym: H-DL-His-OH PubChem CID: 773 ChEBI: CHEBI:27570 IUPAC Name: 2-amino-3-(1H-imidazol-5-yl)propanoic acid SMILES: C1=C(NC=N1)CC(C(=O)O)N