accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO(R)OH(S)(S)O(R)H3C(S)O(S)H3C

CAS RN 50-04-4

IUPAC Name: 2-[(1R,3aS,3bS,9aR,9bS,11aS)-1-hydroxy-9a,11a-dimethyl-7,10-dioxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonyms: cortisone acetate biocort acetate cortisone acetate preparation cortadren irisone acetate cortogen acetate ricortex cortisone acetate (substance) corlin cortelan
Molecular Weight (g/mol): 402.49
Molecular Formula: C23H30O6
InChi Key: ITRJWOMZKQRYTA-RFZYENFJSA-N
SMILES: CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)