Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cortisone Acetate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014337 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
300 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4337 300 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4337 Supplier MilliporeSigma Supelco Supplier No. PHR1923300MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

TRUSTED_SUSTAINABILITY

Specifications

CAS 50-04-4
Linear Formula C23H30O6
MDL Number MFCD00003609
Synonym Cortisone acetate; 17 alpha,21-Dihydroxy-4-pregnene-3,11,20-trione 21-acetate; 21-Acetoxy-4-pregnen-17 alpha-ol-3,11,20-trione; 4-Pregnene-17 alpha,21-diol-3,11,20-trione 21-acetate; Cortisone 21-acetate
Type Certified Reference Material
Quantity 300 mg
Formula Weight 402.48
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.