CAS RN 50-12-4
CAS RN 50-12-4
IUPAC Name:
5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Synonyms:
mephenytoin
Molecular Weight (g/mol):
218.26
Molecular Formula:
C12H14N2O2
InChi Key:
GMHKMTDVRCWUDX-UHFFFAOYNA-N
SMILES:
CCC1(NC(=O)N(C)C1=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Mephenytoin
CAS: 50-12-4 Molecular Formula: C12H14N2O2