accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50-34-0

BrH3CCH3NH3COOOCH3CH3

CAS RN 50-34-0

IUPAC Name: methylbis(propan-2-yl)[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium bromide
Synonyms: propantheline bromide
Molecular Weight (g/mol): 448.4
Molecular Formula: C23H30BrNO3
InChi Key: XLBIBBZXLMYSFF-UHFFFAOYSA-M
SMILES: [Br-].CC(C)[N+](C)(CCOC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12)C(C)C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results