Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propantheline bromide, 99.72%, MP Biomedicals™

Catalog No. ICN15639280 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15639280 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15639280 Supplier MP Biomedicals, Inc Supplier No. 0215639280

missing translation for 'validMainframeMsg'

Specifications

CAS 50-34-0
Color White to White-Yellow
Molecular Formula C23H30BrNO3
Quantity 100 g
Synonym propantheline bromide, pro-banthine, neometantyl, neopepulsan, corrigast, ercorax, ercotina, ketaman, kivatin
InChI Key XLBIBBZXLMYSFF-UHFFFAOYSA-M
SMILES CC(C)[N+](C)(CCOC(=O)C1C2=CC=CC=C2OC3=CC=CC=C13)C(C)C.[Br-]
IUPAC Name methyl-di(propan-2-yl)-[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium;bromide
Molecular Weight (g/mol) 448.401
PubChem CID 9279
Formula Weight 448.4
Percent Purity 0.9972
Physical Form Powder
Chemical Name or Material Propantheline bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.