CAS RN 50-36-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CERILLIANT™ Cocaine Solution, 1.0 mg/mL in acetonitrile, Sold by MilliporeSigma™ Supelco™
A Certified Spiking Solution™ suitable for use in LC/MS or GC/MS testing applications for forensic analysis, urine drug testing, or clinical toxicology.