accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOClClCl

CAS RN 50-73-7

IUPAC Name: 2,3,5-trichlorobenzoate
Synonyms: 2,3,5-trichlorobenzoate
Molecular Weight (g/mol): 224.44
Molecular Formula: C7H2Cl3O2
InChi Key: CGFDSIZRJWMQPP-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Cl)=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results