accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 50-90-8

IUPAC Name: 5-chloro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-chloro-2'-deoxyuridine
Molecular Weight (g/mol): 262.65
Molecular Formula: C9H11ClN2O5
InChi Key: NJCXGFKPQSFZIB-RRKCRQDMSA-N
SMILES: OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(Cl)C(=O)NC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results