Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2'-deoxyuridine, MP Biomedicals™

Catalog No. ICN10547883 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10547883 0.25 g
ICN10547880 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10547883 Supplier MP Biomedicals, Inc Supplier No. 0210547883
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 50-90-8
Molecular Formula C9H11ClN2O5
Molecular Weight (g/mol) 262.646
InChI Key NJCXGFKPQSFZIB-XVMARJQXSA-N
Synonym 5-chloro-2'-deoxyuridine
PubChem CID 13831089
IUPAC Name 5-chloro-1-[(2S,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES C1C(C(OC1N2C=C(C(=O)NC2=O)Cl)CO)O

Specifications

Color White
Assay Percent Range >98%
Quantity 0.25 g
Formula Weight 262.65
Physical Form Powder
Chemical Name or Material 5-Chloro-2'-deoxyuridine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.