accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5000-46-4

CH3OSCH3OO

CAS RN 5000-46-4

IUPAC Name: 1-methanesulfonylpropan-2-one
Synonyms: 1-methanesulfonylpropan-2-one
Molecular Weight (g/mol): 136.17
Molecular Formula: C4H8O3S
InChi Key: NWEYGXQKFVGUFR-UHFFFAOYSA-N
SMILES: CC(=O)CS(C)(=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)