accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5000-48-6

IUPAC Name: 1-(4-chlorobenzenesulfonyl)propan-2-one
Synonyms: 1-(4-chlorobenzenesulfonyl)propan-2-one
Molecular Weight (g/mol): 232.68
Molecular Formula: C9H9ClO3S
InChi Key: BRDBHPZILGTBFY-UHFFFAOYSA-N
SMILES: CC(=O)CS(=O)(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results