CAS RN 500004-32-0
IUPAC Name:
1-(pyren-1-yl)propan-1-one
Synonyms:
1-(pyren-1-yl)propan-1-one
Molecular Weight (g/mol):
258.32
Molecular Formula:
C19H14O
InChi Key:
COIWAOXGROSMQJ-UHFFFAOYSA-N
SMILES:
CCC(=O)C1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
1
–
1
of
1
results
1
Promotions Available
1-Propionylpyrene, 98%, Thermo Scientific™
1-Propionylpyrene, 98%, Thermo Scientific™
CAS: 500004-32-0 Molecular Formula: C19H14O Molecular Weight (g/mol): 258.31 InChI Key: COIWAOXGROSMQJ-UHFFFAOYSA-N PubChem CID: 232861 IUPAC Name: 1-pyren-1-ylpropan-1-one SMILES: CCC(=O)C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1