accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 500790-38-5

IUPAC Name: 3-nitro-N,N-dipropylbenzamide
Synonyms: 3-nitro-N,N-dipropylbenzamide
Molecular Weight (g/mol): 250.3
Molecular Formula: C13H18N2O3
InChi Key: JIPRMVHOCIYIKR-UHFFFAOYSA-N
SMILES: CCCN(CCC)C(=O)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results