accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 501427-92-5

IUPAC Name: 5-acetamido-2-[(2-{[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl)oxy]-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Synonyms: 5-acetamido-2-[(2-{[5-acetamido-4-hydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl)oxy]-4-hydroxy-6-(1,2,3-trihydroxypropyl)oxane-2-carboxylic acid
Molecular Weight (g/mol): 795.7
Molecular Formula: C31H45N3O21
InChi Key: OUJZEVYPUZNYBL-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(O)C(OC2OC(CO)C(O)C(OC3(CC(O)C(NC(C)=O)C(O3)C(O)C(O)CO)C(O)=O)C2O)C(CO)OC1OC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results