CAS RN 501437-28-1
CAS RN 501437-28-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ RSK Inhibitor II
The RSK Inhibitor II, also referenced under CAS 501437-28-1, controls the biological activity of RSK. This small molecule/inhibitor is primarily used for Cell Signaling applications.