accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 5018-30-4

IUPAC Name: 1,3-dimethyl 2-methoxypropanedioate
Synonyms: 1,3-dimethyl 2-methoxypropanedioate
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: ORXJMBXYSGGCHG-UHFFFAOYSA-N
SMILES: COC(C(=O)OC)C(=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results