Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Methoxymalonate 95.0+%, TCI America™
SDP

Catalog No. M08365G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08365G 5 g
M083625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08365G Supplier TCI America Supplier No. M08365G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5018-30-4
Molecular Formula C6H10O5
Molecular Weight (g/mol) 162.14
MDL Number MFCD00009847
InChI Key ORXJMBXYSGGCHG-UHFFFAOYSA-N
Synonym Methoxymalonic Acid Dimethyl Ester
PubChem CID 78718
IUPAC Name 1,3-dimethyl 2-methoxypropanedioate
SMILES COC(C(=O)OC)C(=O)OC

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 162.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Methoxymalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.