accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 5019-82-9

IUPAC Name: bicyclo[3.2.1]octan-2-one
Synonyms: bicyclo[3.2.1]octan-2-one
Molecular Weight (g/mol): 124.18
Molecular Formula: C8H12O
InChi Key: SXFPXZSENHPCSH-UHFFFAOYNA-N
SMILES: O=C1CCC2CCC1C2
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)