accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH

CAS RN 502-41-0

IUPAC Name: cycloheptanol
Synonyms: cycloheptanol
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: QCRFMSUKWRQZEM-UHFFFAOYSA-N
SMILES: OC1CCCCCC1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)