Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cycloheptanol 99%, Thermo Scientific™

Catalog No. AC111031000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC111031000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111031000 Supplier Thermo Scientific Chemicals Supplier No. 111031000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-41-0
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00004150
InChI Key QCRFMSUKWRQZEM-UHFFFAOYSA-N
Synonym suberol, hydroxycycloheptane, 1-cycloheptanol, suberyl alcohol, cycloheptanol, acmc-209kko
PubChem CID 10399
IUPAC Name cycloheptanol
SMILES C1CCCC(CC1)O

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4760 to 1.4780 (20°C, 589nm)
Linear Formula C7H13OH
Quantity 100 g
Beilstein 06, 10
Formula Weight 114.19
Percent Purity ≥98.5%
Physical Form Viscous Liquid
Chemical Name or Material Cycloheptanol, 99%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.