accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 502-73-8

IUPAC Name: hentriacontan-16-one
Synonyms: palmitone 16-hebtriacontanone hebtriacontanone hentricontan-16-one
Molecular Weight (g/mol): 450.84
Molecular Formula: C31H62O
InChi Key: UNRFDARCMOHDBJ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results