Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

16-Hentriacontanone 95.0+%, TCI America™
SDP

Catalog No. H042325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H042325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H042325G Supplier TCI America Supplier No. H042325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502-73-8
Molecular Formula C31H62O
Molecular Weight (g/mol) 450.836
MDL Number MFCD00059222
InChI Key UNRFDARCMOHDBJ-UHFFFAOYSA-N
Synonym Dipentadecyl Ketone, Palmitone
PubChem CID 94741
ChEBI CHEBI:5658
IUPAC Name hentriacontan-16-one
SMILES CCCCCCCCCCCCCCCC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 25 g
Formula Weight 450.84
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 16-Hentriacontanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.