accessibility menu, dialog, popup

UserName

Viewing profile as user:
NI

CAS RN 502161-03-7

IUPAC Name: 3-iodo-9-phenyl-9H-carbazole
Synonyms: 3-iodo-9-phenylcarbazole
Molecular Weight (g/mol): 369.21
Molecular Formula: C18H12IN
InChi Key: PJUAIXDOXUXBDR-UHFFFAOYSA-N
SMILES: IC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results