Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodo-9-phenylcarbazole 98.0+%, TCI America™
SDP

Catalog No. I09135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09135G 5 g
I09131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09135G Supplier TCI America Supplier No. I09135G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502161-03-7
Molecular Formula C18H12IN
Molecular Weight (g/mol) 369.205
MDL Number MFCD09261276
InChI Key PJUAIXDOXUXBDR-UHFFFAOYSA-N
Synonym 3-iodo-n-phenylcarbazole, 3-iodo-9-phenyl-9h-carbazole, 9h-carbazole, 3-iodo-9-phenyl, 3-iodo-9-phenyl-carbazole, 3-iodo-n-phenyl-carbazole, ksc493m9j, 3-iodo-9-phenyl-9h-carbazol
PubChem CID 11810740
IUPAC Name 3-iodo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=CC=CC=C42

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 5 g
Formula Weight 369.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Iodo-9-phenylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.