accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 502169-73-5

IUPAC Name: lithium(1+) tetrabromogolduide
Synonyms: lithium(1+) tetrabromoaurate(III)
Molecular Weight (g/mol): 523.52
Molecular Formula: AuBr4Li
InChi Key: TXCYZTOOKNAEMQ-UHFFFAOYSA-J
SMILES: [Li+].Br[Au-](Br)(Br)Br
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)