Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lithium tetrachloroaurate(III) hydrate, Premion™, 99.99% (metals basis), Thermo Scientific™

Catalog No. AA4042903
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4042903 1 g
AA4042901 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4042903 Supplier Thermo Scientific Chemicals Supplier No. 4042903
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502169-73-5
MDL Number MFCD00798533
Synonym lithium tetrachloroaurate 1-, lithium tetrachloroaurate iii, lithium tetrachloroaurate iii hydrate, lithium 1+ ion tetrachloroaurate
PubChem CID 16217372

Specifications

Physical Form Crystalline, Liquid
Quantity 1 g
Molecular Formula AuBr4Li
InChI Key TXCYZTOOKNAEMQ-UHFFFAOYSA-J
SMILES [Li+].Br[Au-](Br)(Br)Br
IUPAC Name lithium(1+) tetrabromogolduide
Molecular Weight (g/mol) 523.52
Formula Weight 345.72 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Lithium tetrachloroaurate(III) hydrate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.