accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 502481-30-3

IUPAC Name: sodium 2-amino-3-(4-hydroxy-3,5-dinitrophenyl)propanoate
Synonyms: sodium 2-amino-3-(4-hydroxy-3,5-dinitrophenyl)propanoate
Molecular Weight (g/mol): 293.17
Molecular Formula: C9H8N3NaO7
InChi Key: SDPPRYPYKFDRGM-UHFFFAOYNA-M
SMILES: [Na+].NC(CC1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results