Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitro-L-tyrosine Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. D33791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33791G Supplier TCI America Supplier No. D33791G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 502481-30-3
Molecular Formula C9H8N3NaO7
Molecular Weight (g/mol) 293.167
InChI Key SDPPRYPYKFDRGM-JEDNCBNOSA-M
PubChem CID 44629792
IUPAC Name sodium;(2S)-2-amino-3-(4-hydroxy-3,5-dinitrophenyl)propanoate
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])O)[N+](=O)[O-])CC(C(=O)[O-])N.[Na+]

Specifications

Quantity 1 g
Formula Weight 293.17
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dinitro-L-tyrosine Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.