accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNOOBrBrOH3C

CAS RN 502639-02-3

IUPAC Name: 4,5-dibromo-N-methoxy-N-methylfuran-2-carboxamide
Synonyms: 4,5-dibromo-N-methoxy-N-methylfuran-2-carboxamide
Molecular Weight (g/mol): 312.95
Molecular Formula: C7H7Br2NO3
InChi Key: NHVQYVKJJONFAB-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC(Br)=C(Br)O1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results