Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dibromo-N-methoxy-N-methyl-2-furancarboxamide 96.0+%, TCI America™
SDP

Catalog No. D29731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29731G Supplier TCI America Supplier No. D29731G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 502639-02-3
Molecular Formula C7H7Br2NO3
Molecular Weight (g/mol) 312.95
MDL Number MFCD08276337
InChI Key NHVQYVKJJONFAB-UHFFFAOYSA-N
Synonym Methyl 4,5-Dibromo-N-methyl-2-furohydroxamate
PubChem CID 44629754
IUPAC Name 4,5-dibromo-N-methoxy-N-methylfuran-2-carboxamide
SMILES CON(C)C(=O)C1=CC(Br)=C(Br)O1

Specifications

Quantity 1 g
Formula Weight 312.95
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dibromo-N-methoxy-N-methyl-2-furancarboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.