accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 503-38-8

IUPAC Name: trichloromethyl carbonochloridate
Synonyms: diphosgene
Molecular Weight (g/mol): 197.82
Molecular Formula: C2Cl4O2
InChi Key: HCUYBXPSSCRKRF-UHFFFAOYSA-N
SMILES: ClC(=O)OC(Cl)(Cl)Cl
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)