Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trichloromethyl Chloroformate

Catalog No. 2326150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-261-50ML 50 mL
23-261-10ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 23-261-50ML Supplier Sigma Aldrich Supplier No. 2326150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Trichloromethyl chloroformate (TCF) is an effective alternative to phosgene. It can react with amines, amino acids and amino alcohols to give the corresponding isocyanates, isocyanato acid chlorides and isocyanato chloroformates. TCF can also be used: To synthesize N-carboxy +a-amino acid anhydrides.As an acylating agent to synthesize oxazolidinones from +a-amino alcohols.As a dehydrating agent to synthesize aromatic diisocyanides in the presence of triethylamine.Easy to handle substitute for phosgene; In-situ charcoal-catalyzed decomposition to phosgene and reaction with amino acids to N-carboxy anhydrides

Specifications

CAS 503-38-8
Density 1.639 g/mL at 20°C
Boiling Point 20°C (10 mmHg, lit.)
Molecular Formula C2Cl4O2
Refractive Index n20/D 1.458
Linear Formula ClCOOCCl3
MDL Number MFCD00015553
Quantity 50 mL
Synonym TCF; Diphosgene; di-Phosgene
Molecular Weight (g/mol) 197.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.