accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 503-93-5

IUPAC Name: 2,6,6-trimethylcyclohepta-2,4-dien-1-one
Synonyms: 2,6,6-trimethylcyclohepta-2,4-dien-1-one
Molecular Weight (g/mol): 150.22
Molecular Formula: C10H14O
InChi Key: QNGQIURXCUHNAT-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)(C)CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results